Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-1,2-Propanediol, 97%

Click to view available options
Quantity:
1g
5g
25g
4254-15-3
C3H8O2
76.095
MFCD00004539
DNIAPMSPPWPWGF-VKHMYHEASA-N
s-+-1,2-propanediol,s-propane-1,2-diol,s-1,2-propanediol,2s-propane-1,2-diol,s-propylene glycol,s-+-propylene glycol,s-+-1,2-dihydroxypropane,propylene glycol #,pgo
439846
CHEBI:29002
(2S)-propane-1,2-diol
CC(CO)O
4254-15-3
C3H8O2
76.095
MFCD00004539
DNIAPMSPPWPWGF-VKHMYHEASA-N
s-+-1,2-propanediol,s-propane-1,2-diol,s-1,2-propanediol,2s-propane-1,2-diol,s-propylene glycol,s-+-propylene glycol,s-+-1,2-dihydroxypropane,propylene glycol #,pgo
439846
CHEBI:29002
(2S)-propane-1,2-diol
CC(CO)O
Applications
(S)-(+)-1,2-Propanediol acts as an organic solvent and diluent used in pharmaceutical preparations. It is also used as a chiral synthetic intermediate and substrate for enzyme studies.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides, oxidizing agents,chloroformates and reducing agents.
Optical Rotation +16° (Neat)
Boiling Point 186°C to 188°C
Flash Point 107°C (224°F)
Quantity 1g
Beilstein 1718871
Merck Index 14,7855
Formula Weight 76.1
Percent Purity 97%
Chemical Name or Material (S)-(+)-1,2-Propanediol