Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1,2-Propanediol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
Packaging:
Glass bottle
4254-15-3
C3H8O2
76.095
MFCD00004539
DNIAPMSPPWPWGF-VKHMYHEASA-N
s-+-1,2-propanediol,s-propane-1,2-diol,s-1,2-propanediol,2s-propane-1,2-diol,s-propylene glycol,s-+-propylene glycol,s-+-1,2-dihydroxypropane,propylene glycol #,pgo
439846
CHEBI:29002
(2S)-propane-1,2-diol
CC(CO)O
4254-15-3
C3H8O2
76.095
MFCD00004539
DNIAPMSPPWPWGF-VKHMYHEASA-N
s-+-1,2-propanediol,s-propane-1,2-diol,s-1,2-propanediol,2s-propane-1,2-diol,s-propylene glycol,s-+-propylene glycol,s-+-1,2-dihydroxypropane,propylene glycol #,pgo
439846
CHEBI:29002
(2S)-propane-1,2-diol
CC(CO)O
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Density 102g/mL
Quantity 1g
Specific Rotation Condition + 16.00 (20.00°C c=neat)
Refractive Index 1.4310 to 1.4350 (20°C, 589nm)
Specific Rotation +15° to +18° (20°C, 589nm) (neat)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH(OH)CH2OH
Merck Index 15, 7968
Formula Weight 76.1
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material (S)-(+)-1, 2-Propanediol
Show More Show Less