Alfa Aesar™ (S)-1-(3-Bromophenyl)ethylamine, ChiPros™ 99%, ee 98+%

Manufacturer: Alfa Aesar™ H2699806

Quantity 5g

 View more versions of this product

Catalog No. AAH2699806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 139305-96-7
    Molecular Formula C8H10BrN
    Formula Weight 200.08
    MDL Number MFCD00671638
    UN Number UN2735
    Synonym (S)-m-Bromo-alpha-methylbenzylamine
    Chemical Name or Material (S)-1-(3-Bromophenyl)ethylamine

    Specifications

    Formula Weight 200.08
    Optical Rotation −22° (Neat)
    Molecular Formula C8H10BrN
    MDL Number MFCD00671638
    CAS 139305-96-7
    UN Number UN2735
    Synonym (S)-m-Bromo-alpha-methylbenzylamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Boiling Point 96°C (4mmHg)
    Quantity 5g
    Flash Point >110°C (230°F)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (S)-1-(3-Bromophenyl)ethylamine
    Assay 99%, ee ≥98%

    Description

    Applications
    (S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents, carbon monoxide and strong acids.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit