Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-(4-Chlorophenyl)ethylamine, prd by BASF AG, 95+% (sum of isomers), 95% e., Acros Organics

Click to view available options
Quantity:
5g
4187-56-8
C8H10ClN
155.625
MFCD00671640
PINPOEWMCLFRRB-LURJTMIESA-N
s-1-4-chlorophenyl ethylamine,s-1-4-chlorophenyl ethanamine,1s-1-4-chlorophenyl ethanamine,s-4-chloro-alpha-methylbenzylamine,s---1-4-chlorophenyl ethylamine,1s-1-4-chlorophenyl ethan-1-amine,pubchem15221,s-4-chloro-,a-methylbenzylamine,1s-1-4-chlorophenyl ethylamine,s-1-4-chloro-phenyl ethylamine
1550852
(1S)-1-(4-chlorophenyl)ethanamine
CC(C1=CC=C(C=C1)Cl)N
4187-56-8
C8H10ClN
155.625
MFCD00671640
PINPOEWMCLFRRB-LURJTMIESA-N
s-1-4-chlorophenyl ethylamine,s-1-4-chlorophenyl ethanamine,1s-1-4-chlorophenyl ethanamine,s-4-chloro-alpha-methylbenzylamine,s---1-4-chlorophenyl ethylamine,1s-1-4-chlorophenyl ethan-1-amine,pubchem15221,s-4-chloro-,a-methylbenzylamine,1s-1-4-chlorophenyl ethylamine,s-1-4-chloro-phenyl ethylamine
1550852
(1S)-1-(4-chlorophenyl)ethanamine
CC(C1=CC=C(C=C1)Cl)N
Enantiomeric Excess 95.0% min.
Specific Rotation -22° to -25° (20°C, 589nm) (C=2, methanol)
Refractive Index 1.5410 to 1.5430 (20°C, 589nm)
Specific Rotation Condition - 23.50 (20.00°C c=2,methanol)
Infrared Spectrum Authentic
Density 1.1150g/mL
Quantity 5g
Color Colorless to Yellow
Linear Formula ClC6H4CH(CH3)NH2
Formula Weight 155.63
Physical Form Liquid
Percent Purity ≥95.0% sum of isomers
Chemical Name or Material (S)-1-(4-Chlorophenyl)ethylamine, produced by BASF AG
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger