Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
93643-24-4
C7H14N2
126.2
FTTATHOUSOIFOQ-ZETCQYMHSA-N
s-octahydropyrrolo 1,2-a pyrazine,s-1,4-diazabicyclo 4.3.0 nonane,8as-octahydropyrrolo 1,2-a pyrazine,8as-octahydropyrrolo 1,2-a piperazine,s-octahydro-pyrrolo 1,2-a pyrazine,6s-1,4-diazabicyclo 4.3.0 nonane,pyrrolo 1,2-a pyrazine, octahydro-, 8as,8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine,pubchem20597,6s-1,4-diazabicyclo 4,3,0 nonane
781249
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
C1CC2CNCCN2C1
93643-24-4
C7H14N2
126.2
FTTATHOUSOIFOQ-ZETCQYMHSA-N
s-octahydropyrrolo 1,2-a pyrazine,s-1,4-diazabicyclo 4.3.0 nonane,8as-octahydropyrrolo 1,2-a pyrazine,8as-octahydropyrrolo 1,2-a piperazine,s-octahydro-pyrrolo 1,2-a pyrazine,6s-1,4-diazabicyclo 4.3.0 nonane,pyrrolo 1,2-a pyrazine, octahydro-, 8as,8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine,pubchem20597,6s-1,4-diazabicyclo 4,3,0 nonane
781249
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
C1CC2CNCCN2C1
Specific Rotation Condition + 5.88 (20.00°C c=1, CHCl3)
Quantity 1g
Infrared Spectrum Authentic
Specific Rotation + 5.88
Flash Point 89°C
Assay Percent Range 97.5% min. (GC)
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger