Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Benzyl-3-aminopyrrolidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
114715-38-7
C11H16N2
176.26
HBVNLKQGRZPGRP-NSHDSACASA-N
s-+-1-benzyl-3-aminopyrrolidine,3s-1-benzylpyrrolidin-3-amine,3s-+-1-benzyl-3-aminopyrrolidine,s-n-benzyl-3-aminopyrrolidine,s-3-amino-1-n-benzyl-pyrrolidine,s-1-benzylpyrrolidin-3-amine,s-1-benzyl-3-aminopyrrolidine,s-3-amino-1-benzylpyrrolidine,s-bap,s-3-amino-1-n-benzylpyrrolidine
1519353
(3S)-1-benzylpyrrolidin-3-amine
C1CN(CC1N)CC2=CC=CC=C2
114715-38-7
C11H16N2
176.26
HBVNLKQGRZPGRP-NSHDSACASA-N
s-+-1-benzyl-3-aminopyrrolidine,3s-1-benzylpyrrolidin-3-amine,3s-+-1-benzyl-3-aminopyrrolidine,s-n-benzyl-3-aminopyrrolidine,s-3-amino-1-n-benzyl-pyrrolidine,s-1-benzylpyrrolidin-3-amine,s-1-benzyl-3-aminopyrrolidine,s-3-amino-1-benzylpyrrolidine,s-bap,s-3-amino-1-n-benzylpyrrolidine
1519353
(3S)-1-benzylpyrrolidin-3-amine
C1CN(CC1N)CC2=CC=CC=C2
Boiling Point 108°C to 115°C (5.0mmHg)
Flash Point 140°C
Specific Gravity 1.02
Density 1.0200g/mL
Specific Rotation + 2
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 176.26
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(+)-1-Benzyl-3-aminopyrrolidine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger