Alfa Aesar™ (S)-(+)-1-Benzyl-3-hydroxypiperidine, 97%

Manufacturer: Alfa Aesar™ H3061503

Quantity 1g

 View more versions of this product

Catalog No. AAH3061503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91599-79-0
    Molecular Formula C12H17NO
    Formula Weight 191.27
    MDL Number MFCD03426359
    Synonym (S)-1-Benzyl-3-piperidinol
    Chemical Name or Material (S)-(+)-1-Benzyl-3-hydroxypiperidine

    Specifications

    Formula Weight 191.27
    Molecular Formula C12H17NO
    MDL Number MFCD03426359
    CAS 91599-79-0
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Recommended Storage 2° to 8°C
    Synonym (S)-1-Benzyl-3-piperidinol
    Chemical Name or Material (S)-(+)-1-Benzyl-3-hydroxypiperidine
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit