Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-BOC-3-(Aminomethyl)piperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
140645-24-5
C11H22N2O2
214.31
WPWXYQIMXTUMJB-VIFPVBQESA-N
s-1-boc-3-aminomethyl piperidine,s-n-boc-3-aminomethylpiperidine,s-3-aminomethyl-1-boc-piperidine,s-tert-butyl 3-aminomethyl piperidine-1-carboxylate,s-1-n-boc-3-aminomethyl piperidine,s-3-aminomethyl-1-n-boc-piperidine,tert-butyl 3s-3-aminomethyl piperidine-1-carboxylate,s-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester,s-1-n-boc-piperidine-3-methylamine,3s-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502022
tert-butyl (3S)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN
140645-24-5
C11H22N2O2
214.31
WPWXYQIMXTUMJB-VIFPVBQESA-N
s-1-boc-3-aminomethyl piperidine,s-n-boc-3-aminomethylpiperidine,s-3-aminomethyl-1-boc-piperidine,s-tert-butyl 3-aminomethyl piperidine-1-carboxylate,s-1-n-boc-3-aminomethyl piperidine,s-3-aminomethyl-1-n-boc-piperidine,tert-butyl 3s-3-aminomethyl piperidine-1-carboxylate,s-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester,s-1-n-boc-piperidine-3-methylamine,3s-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502022
tert-butyl (3S)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN
Specific Rotation -15
Color Colorless
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 214.31
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (S)-1-BOC-3-(Aminomethyl)piperidine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning