Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine, 97%

Click to view available options
Quantity:
250mg
1g
199175-10-5
C10H20N2O2
200.282
MFCD02179397
OGCCBDIYOAFOGK-QMMMGPOBSA-N
s-1-boc-3-aminomethyl pyrrolidine,s-3-aminomethyl-1-n-boc-pyrrolidine,tert-butyl 3s-3-aminomethyl pyrrolidine-1-carboxylate,s-tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate,3s-3-aminomethyl-1-boc-pyrrolidine,s-1-n-boc-3-aminomethyl pyrrolidine,s-1-boc-3-aminomethylpyrrolidine,s-3-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester,s-3-aminomethyl-1-boc-pyrrolidine,s-tert-butyl-3-aminomethyl pyrrolidine-1-carboxylate
1514454
tert-butyl (3S)-3-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CN
199175-10-5
C10H20N2O2
200.282
MFCD02179397
OGCCBDIYOAFOGK-QMMMGPOBSA-N
s-1-boc-3-aminomethyl pyrrolidine,s-3-aminomethyl-1-n-boc-pyrrolidine,tert-butyl 3s-3-aminomethyl pyrrolidine-1-carboxylate,s-tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate,3s-3-aminomethyl-1-boc-pyrrolidine,s-1-n-boc-3-aminomethyl pyrrolidine,s-1-boc-3-aminomethylpyrrolidine,s-3-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester,s-3-aminomethyl-1-boc-pyrrolidine,s-tert-butyl-3-aminomethyl pyrrolidine-1-carboxylate
1514454
tert-butyl (3S)-3-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CN
Color Colorless to Yellow
Quantity 1g
Refractive Index 1.4780
Sensitivity Air Sensitive
Formula Weight 200.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine

storageNote1 : 2° to 8°C