Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
1221793-43-6
C21H19NO5
365.385
MFCD03094868
YGAHCGORXONSSM-IBGZPJMESA-N
s-1-fmoc-4-oxopiperidine-2-carboxylic acid,fmoc-l-homopro 4-oxo,2s-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
49759859
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
1221793-43-6
C21H19NO5
365.385
MFCD03094868
YGAHCGORXONSSM-IBGZPJMESA-N
s-1-fmoc-4-oxopiperidine-2-carboxylic acid,fmoc-l-homopro 4-oxo,2s-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
49759859
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Melting Point 228°C to 223°C
Sensitivity Moisture Sensitive
Color White to Yellow
Quantity 1g
Formula Weight 365.38
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid

storageNote1 : 2° to 8°C