Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Methoxy-2-propanol, 99%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol,2s-1-methoxypropan-2-ol,s-1-methoxypropan-2-ol,s---1-methoxy-2-propanol,s-1-methoxy-2-propanol,2-propanol, 1-methoxy-, 2s,2s-1-methoxy-2-propanol,pubchem6715,s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol,2s-1-methoxypropan-2-ol,s-1-methoxypropan-2-ol,s---1-methoxy-2-propanol,s-1-methoxy-2-propanol,2-propanol, 1-methoxy-, 2s,2s-1-methoxy-2-propanol,pubchem6715,s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O
Quantity 1mL
Infrared Spectrum Authentic
Flash Point 30°C
Packaging Glass bottle
Color Colorless
Melting Point -97.0°C
Density 0.9200g/mL
Boiling Point 120.0°C
Specific Rotation + 20.00
Refractive Index 1.4020 to 1.4040
Specific Gravity 0.92
Specific Rotation Condition +20° (20°C c=10,CHCl3)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3OCH2CH(OH)CH3
Solubility Information Solubility in water: soluble
Formula Weight 90.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(+)-1-Methoxy-2-propanol
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning