Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Methoxy-2-propanol, 99%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol,2s-1-methoxypropan-2-ol,s-1-methoxypropan-2-ol,s---1-methoxy-2-propanol,s-1-methoxy-2-propanol,2-propanol, 1-methoxy-, 2s,2s-1-methoxy-2-propanol,pubchem6715,s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol,2s-1-methoxypropan-2-ol,s-1-methoxypropan-2-ol,s---1-methoxy-2-propanol,s-1-methoxy-2-propanol,2-propanol, 1-methoxy-, 2s,2s-1-methoxy-2-propanol,pubchem6715,s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O
Boiling Point 120.0°C
Quantity 25mL
Flash Point 30°C
Specific Gravity 0.92
Infrared Spectrum Authentic
Specific Rotation Condition +20° (20°C c=10,CHCl3)
Packaging Glass bottle
Color Colorless
Density 0.9200g/mL
Refractive Index 1.4020 to 1.4040
Melting Point -97.0°C
Specific Rotation + 20.00
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OCH2CH(OH)CH3
Solubility Information Solubility in water: soluble
Formula Weight 90.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(+)-1-Methoxy-2-propanol
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning