Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
104641-59-0
C5H11NO
101.15
FLVFPAIGVBQGET-YFKPBYRVSA-N
s-+-1-methyl-3-pyrrolidinol,3s-1-methylpyrrolidin-3-ol,s-1-methyl-3-pyrrolidinol,s-1-methylpyrrolidin-3-ol,3-pyrrolidinol, 1-methyl-, 3s,s-+-3-hydroxy-n-methylpyrrolidine,s-+-1-methyl-3-hydroxypyrrolidine,s-3-hydroxy-1-methylpyrrolidine,1-methyl-3-pyrrolidinol, +,s-3-hydroxy-1-methyl-pyrrolidine
643504
(3S)-1-methylpyrrolidin-3-ol
CN1CCC(C1)O
104641-59-0
C5H11NO
101.15
FLVFPAIGVBQGET-YFKPBYRVSA-N
s-+-1-methyl-3-pyrrolidinol,3s-1-methylpyrrolidin-3-ol,s-1-methyl-3-pyrrolidinol,s-1-methylpyrrolidin-3-ol,3-pyrrolidinol, 1-methyl-, 3s,s-+-3-hydroxy-n-methylpyrrolidine,s-+-1-methyl-3-hydroxypyrrolidine,s-3-hydroxy-1-methylpyrrolidine,1-methyl-3-pyrrolidinol, +,s-3-hydroxy-1-methyl-pyrrolidine
643504
(3S)-1-methylpyrrolidin-3-ol
CN1CCC(C1)O
Specific Gravity 0.993
Flash Point 90°C
Boiling Point 181°C to 182°C
Specific Rotation 7
Water 2% Max.
Density 0.9930g/mL
Quantity 1g
Infrared Spectrum Authentic
Color Brown to Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 101.15
Physical Form Liquid
Percent Purity 98%,ee99%
Chemical Name or Material (S)-(+)-1-Methyl-3-pyrrolidinol, ee 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : May darken on storage with no loss of purity