Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-(p-Tolyl)ethylamine 98.0+%, TCI America™

Click to view available options
Quantity:
1mL
5mL
27298-98-2
C9H13N
135.21
MFCD00145246
UZDDXUMOXKDXNE-QMMMGPOBSA-N
s-1-p-tolyl ethanamine,s---1-p-tolyl ethylamine,s---4-1-aminoethyl toluene,s-1-p-tolyl ethylamine,1s-1-4-methylphenyl ethan-1-amine,1s-1-p-tolyl ethanamine,1s-1-4-methylphenyl ethanamine,s-1-p-tolylethanamine,1-4-methylphenyl ethanamine #,s-1-4-methylphenyl ethylamine
7015759
(1S)-1-(4-methylphenyl)ethanamine
CC1=CC=C(C=C1)C(C)N
27298-98-2
C9H13N
135.21
MFCD00145246
UZDDXUMOXKDXNE-QMMMGPOBSA-N
s-1-p-tolyl ethanamine,s---1-p-tolyl ethylamine,s---4-1-aminoethyl toluene,s-1-p-tolyl ethylamine,1s-1-4-methylphenyl ethan-1-amine,1s-1-p-tolyl ethanamine,1s-1-4-methylphenyl ethanamine,s-1-p-tolylethanamine,1-4-methylphenyl ethanamine #,s-1-4-methylphenyl ethylamine
7015759
(1S)-1-(4-methylphenyl)ethanamine
CC1=CC=C(C=C1)C(C)N
Quantity 5mL
Color Yellow
Boiling Point 105°C
UN Number 2735
Formula Weight 135.21
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material (S)-(-)-1-(p-Tolyl)ethylamine