Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Phenyl-1,2-ethanediol, 99+%, Thermo Scientific™

Catalog No. AC264640010
Click to view available options
:
1g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25779-13-9
CAS Min % 99.0
CAS Max % 100.0
Boiling Point 272°C to 274°C (755.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H10O2
Quantity 1g
MDL Number MFCD00066256
Merck Index 14, 8861
Specific Rotation Condition +59.5° (20°C c=2,1,2-dichloroEt)
Specific Rotation + 59.5
Synonym s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,ether,chloroform
InChI Key PWMWNFMRSKOCEY-MRVPVSSYSA-N
SMILES C1=CC=C(C=C1)C(CO)O
IUPAC Name (1S)-1-phenylethane-1,2-diol
Molecular Weight (g/mol) 138.16
PubChem CID 643312
Formula Weight 138.16
Percent Purity 99+%
Chemical Name or Material (S)-(+)-1-Phenyl-1, 2-ethanediol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.