Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(s)-(-)-1-phenyl-1-propanol, 95%, Acros Organics

Click to view available options
Quantity:
1g
613-87-6
C9H12O
136.194
MFCD00066207
DYUQAZSOFZSPHD-VIFPVBQESA-N
s---1-phenyl-1-propanol,s-1-phenylpropan-1-ol,1s-1-phenylpropan-1-ol,s-1-phenyl-1-propanol,s---1-phenylpropanol,s-1-phenyl-propan-1-ol,s---,a-ethylbenzyl alcohol,--s-1-phenyl-1-propanol;,s---alpha-ethylbenzyl alcohol
2734864
(1S)-1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O
613-87-6
C9H12O
136.194
MFCD00066207
DYUQAZSOFZSPHD-VIFPVBQESA-N
s---1-phenyl-1-propanol,s-1-phenylpropan-1-ol,1s-1-phenylpropan-1-ol,s-1-phenyl-1-propanol,s---1-phenylpropanol,s-1-phenyl-propan-1-ol,s---,a-ethylbenzyl alcohol,--s-1-phenyl-1-propanol;,s---alpha-ethylbenzyl alcohol
2734864
(1S)-1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O
Color Colorless
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Infrared Spectrum Authentic
Density 0.993g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C2H5CH(OH)C6H5
Formula Weight 136.19
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (S)-(-)-1-Phenyl-1-propanol
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.