Alfa Aesar™ (S)-1-Phenylbutylamine, ChiPros™, 98%, ee 98+%

Manufacturer: Alfa Aesar™ H2704806

Quantity 5g

 View more versions of this product

Catalog No. AAH2704806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3789-60-4
    Molecular Formula C10H15N
    Formula Weight 149.24
    MDL Number MFCD08064291
    UN Number UN2735
    Synonym (S)-1-Amino-1-phenylbutane; (S)-alpha-n-Propylbenzylamine
    Chemical Name or Material (S)-1-Phenylbutylamine

    Specifications

    Formula Weight 149.24
    MDL Number MFCD08064291
    UN Number UN2735
    Synonym (S)-1-Amino-1-phenylbutane; (S)-alpha-n-Propylbenzylamine
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Molecular Formula C10H15N
    CAS 3789-60-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 220°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (S)-1-Phenylbutylamine
    Assay 98%, ee ≥98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit