Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-Phenylethylamine, 99+%, produced by BASF AG, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
2627-86-3
C8H11N
121.18
MFCD00064406
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N
2627-86-3
C8H11N
121.18
MFCD00064406
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N
Chiral derivatization reagent
Water 0.5% max.
Refractive Index 1.5250 to 1.5270
Specific Rotation − 40.00
Specific Gravity 0.94
Specific Rotation Condition −40.00 (20.00°C neat)
Packaging Glass bottle
Quantity 50g
Flash Point 71°C
Density 0.9400g/mL
Color Colorless
Boiling Point 187.0°C to 189.0°C
Melting Point -10.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Linear Formula C6H5CH(CH3)NH2
Beilstein 12,II,587
Fieser 09,363; 13,185; 17,276; 12,319; 11,411; 05,441; 03,199; 01,838; 02,271; 08,393
Merck Index 13, 6054
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Formula Weight 121.18
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 220-098-