Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-Phenylethylamine, 99+%, produced by BASF AG, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
2627-86-3
C8H11N
121.18
MFCD00064406
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N
2627-86-3
C8H11N
121.18
MFCD00064406
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N
Chiral derivatization reagent
Quantity 250g
Refractive Index 1.5250 to 1.5270
Water 0.5% max.
Specific Rotation − 40.00
Specific Rotation Condition −40.00 (20.00°C neat)
Specific Gravity 0.94
Melting Point -10.0°C
Boiling Point 187.0°C to 189.0°C
Infrared Spectrum Authentic
Flash Point 71°C
Packaging Glass bottle
Density 0.9400g/mL
Color Colorless
Assay Percent Range 99+%
Linear Formula C6H5CH(CH3)NH2
Beilstein 12,II,587
Fieser 09,363; 13,185; 17,276; 12,319; 11,411; 05,441; 03,199; 01,838; 02,271; 08,393
Merck Index 13, 6054
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger