Alfa Aesar™ (S)-2-(1-Boc-2-pyrrolidinyl)acetic acid, 95%

Manufacturer: Alfa Aesar™ H5217503

Quantity 1g

 View more versions of this product

Catalog No. AAH5217503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56502-01-3
    Molecular Formula C11H19NO4
    Formula Weight 229.28
    MDL Number MFCD01862939
    Beilstein 478306
    Synonym N-Boc-L-beta-homoproline; Boc-beta3-Homopro-OH
    Chemical Name or Material (S)-2-(1-Boc-2-pyrrolidinyl)acetic acid

    Specifications

    Formula Weight 229.28
    Molecular Formula C11H19NO4
    MDL Number MFCD01862939
    CAS 56502-01-3
    Beilstein 478306
    Synonym N-Boc-L-beta-homoproline; Boc-beta3-Homopro-OH
    Quantity 1g
    Chemical Name or Material (S)-2-(1-Boc-2-pyrrolidinyl)acetic acid
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit