Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97%

Click to view available options
Quantity:
100mg
0.25g
1g
5g
76189-56-5
C44H32P2
622.688
MFCD00010805
MUALRAIOVNYAIW-UHFFFAOYSA-N
binap,r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl,2,2'-bis diphenylphosphino-1,1'-binaphthyl,s-binap,s---binap,s---2,2'-bis diphenylphosphino-1,1'-binaphthyl,r-+-binap,r-binap,rac-binap,2,2'-bis diphenylphosphino-1,1'-binaphthalene
634876
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
76189-56-5
C44H32P2
622.688
MFCD00010805
MUALRAIOVNYAIW-UHFFFAOYSA-N
binap,r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl,2,2'-bis diphenylphosphino-1,1'-binaphthyl,s-binap,s---binap,s---2,2'-bis diphenylphosphino-1,1'-binaphthyl,r-+-binap,r-binap,rac-binap,2,2'-bis diphenylphosphino-1,1'-binaphthalene
634876
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
Quantity 0.25g
Optical Rotation −230° (c=0.3 in Toluene)
Melting Point 239°C to 241°C
Beilstein 5321444
Merck Index 14,1223
Formula Weight 622.69
Percent Purity 97%
Chemical Name or Material (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning