Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2,2-Dimethyl-4-(hydroxymethyl)-1,3-dioxolane-±toluenesulfonate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
23735-43-5
C13H18O5S
286.342
MFCD00063234
SRKDUHUULIWXFT-NSHDSACASA-N
s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate,s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate,4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate,s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate,s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate,r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate,pubchem6795,2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #,s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate
2723760
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C
23735-43-5
C13H18O5S
286.342
MFCD00063234
SRKDUHUULIWXFT-NSHDSACASA-N
s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate,s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate,4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate,s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate,s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate,r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate,pubchem6795,2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #,s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate
2723760
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C
Specific Rotation +7.0° to +8.0° (20°C, 589nm) (c=2, DMF)
Color Tan to Yellow
Infrared Spectrum Authentic
Specific Rotation Condition +4.20° (20°C c=10,abs.alcohol)
Quantity 1g
Refractive Index 1.5055 to 1.5075 (20°C, 589nm)
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 286.33
Physical Form Liquid To Solid
Percent Purity ≥96.0%
Chemical Name or Material (S)-(+)-2, 2-Dimethyl-4-(hydroxymethyl)-1, 3-dioxolane-p-toluenesulfonate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.