Alfa Aesar™ (S)-(+)-2,3-O-Isopropylideneglycerol, 98%

Manufacturer: Alfa Aesar™ B2448804

Quantity 2g

 View more versions of this product

Catalog No. AAB2448804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22323-82-6
    Molecular Formula C6H12O3
    Formula Weight 132.18
    MDL Number MFCD00063239
    Beilstein 80118
    Merck Index 14,5213
    Synonym (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol
    Chemical Name or Material (S)-(+)-2,3-O-Isopropylideneglycerol

    Specifications

    Formula Weight 132.18
    MDL Number MFCD00063239
    Beilstein 80118
    Merck Index 14,5213
    Synonym (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 82° to 83°C (14mmHg)
    Quantity 2g
    Flash Point 80°C (176°F)
    Optical Rotation +11.5° (c=5 in Methanol)
    Recommended Storage 2° to 8°C
    Molecular Formula C6H12O3
    CAS 22323-82-6
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material (S)-(+)-2,3-O-Isopropylideneglycerol
    Assay 98%

    Description

    Applications
    (S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

    Solubility
    Miscible with water and chloroform.

    Notes
    Store in a cool place. Incompatible with acids and strong oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit