Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-2,3-O-Isopropylideneglycerol, 98%

Click to view available options
Quantity:
2g
10g
22323-82-6
C6H12O3
132.159
MFCD00063239
RNVYQYLELCKWAN-YFKPBYRVSA-N
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol,s-glycerol acetonide,s-+-1,2-isopropylideneglycerol,4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol,s-2,2-dimethyl-1,3-dioxolane-4-methanol,s-solketal,1,2-isopropylidene-sn-glycerol,s-+-2,3-o-isopropylideneglycerol,1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s,s-2,2-dimethyl-1,3-dioxolan-4-yl methanol
736057
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
CC1(OCC(O1)CO)C
22323-82-6
C6H12O3
132.159
MFCD00063239
RNVYQYLELCKWAN-YFKPBYRVSA-N
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol,s-glycerol acetonide,s-+-1,2-isopropylideneglycerol,4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol,s-2,2-dimethyl-1,3-dioxolane-4-methanol,s-solketal,1,2-isopropylidene-sn-glycerol,s-+-2,3-o-isopropylideneglycerol,1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s,s-2,2-dimethyl-1,3-dioxolan-4-yl methanol
736057
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
CC1(OCC(O1)CO)C
Applications
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
Flash Point 80°C (176°F)
Optical Rotation +11.5° (c=5 in Methanol)
Quantity 10g
Boiling Point 82°C to 83°C (14mmHg)
Beilstein 80118
Merck Index 14,5213
Formula Weight 132.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2,3-O-Isopropylideneglycerol

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C