Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-2,4-Bis(Fmoc-amino)butyric acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
201473-83-8
C34H30N2O6
562.622
MFCD00237004
ALZDIZDLDRWFAC-HKBQPEDESA-N
fmoc-dab fmoc-oh,s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,2s-2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,ambotzfaa1770,fmoc-l-dab fmoc-oh,n-a,n-?-di-fmoc-l-2,4-diaminobutyric acid,n-,a,n-,a-di-fmoc-l-2,4-diaminobutyric acid,2s-2,4-bis 9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,2s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,butanoicacid, 2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
46737279
(2S)-2,4-bis(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
201473-83-8
C34H30N2O6
562.622
MFCD00237004
ALZDIZDLDRWFAC-HKBQPEDESA-N
fmoc-dab fmoc-oh,s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,2s-2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,ambotzfaa1770,fmoc-l-dab fmoc-oh,n-a,n-?-di-fmoc-l-2,4-diaminobutyric acid,n-,a,n-,a-di-fmoc-l-2,4-diaminobutyric acid,2s-2,4-bis 9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,2s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,butanoicacid, 2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
46737279
(2S)-2,4-bis(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
Quantity 250mg
Formula Weight 562.62
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (S)-2,4-Bis(Fmoc-amino)butyric acid

storageNote1 : -30° to -10°C