Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%, Acros Organics

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
78342-42-4
C9H16N2O2
184.24
MFCD00066229
FCFWEOGTZZPCTO-QMMMGPOBSA-N
s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,2s-+-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine,2s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine,pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2s,2s-2,5-dihydro-2-isopropyl-3,6-dimethoxypyrazine,2s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,3s-3-isopropyl-2,5-dimethoxy-3,6-dihydropyrazine,s-2,5-dihydro-3,6-dimethoxy-2-iso-propylpyrazine,2s-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
736065
(2S)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
CC(C)C1C(=NCC(=N1)OC)OC
78342-42-4
C9H16N2O2
184.24
MFCD00066229
FCFWEOGTZZPCTO-QMMMGPOBSA-N
s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,2s-+-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine,2s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine,pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2s,2s-2,5-dihydro-2-isopropyl-3,6-dimethoxypyrazine,2s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine,3s-3-isopropyl-2,5-dimethoxy-3,6-dihydropyrazine,s-2,5-dihydro-3,6-dimethoxy-2-iso-propylpyrazine,2s-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
736065
(2S)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
CC(C)C1C(=NCC(=N1)OC)OC
Specific Gravity 1.028
Density 1.0280g/mL
Specific Rotation +96° to +107° (20°C, 589nm) (C=1, EtOH)
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 1mL
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Packaging Glass bottle
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 184.24
Physical Form Liquid
Percent Purity 97+%
Chemical Name or Material (S)-2, 5-Dihydro-3, 6-dimethoxy-2-isopropylpyrazine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning