Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-2-Amino-1-propanol, 98%

Click to view available options
Quantity:
1g
5g
25g
2758-06-7
C5H13Br2N
246.97
MFCD00064412
OINMNSFDYTYXEQ-UHFFFAOYSA-M
2-bromoethyl trimethylammonium bromide,2-bromo-n,n,n-trimethylethanaminium bromide,2-bromoethyltrimethylammonium bromide,bromcholin,hypercyl,kathesin,btab,btab the plant regulator,bcb the plant regulator,unii-kh1i7ru18l
17689
2-bromoethyl(trimethyl)azanium;bromide
C[N+](C)(C)CCBr.[Br-]
2758-06-7
C5H13Br2N
246.97
MFCD00064412
OINMNSFDYTYXEQ-UHFFFAOYSA-M
2-bromoethyl trimethylammonium bromide,2-bromo-n,n,n-trimethylethanaminium bromide,2-bromoethyltrimethylammonium bromide,bromcholin,hypercyl,kathesin,btab,btab the plant regulator,bcb the plant regulator,unii-kh1i7ru18l
17689
2-bromoethyl(trimethyl)azanium;bromide
C[N+](C)(C)CCBr.[Br-]
Flash Point 83°C (181°F)
Quantity 25g
Optical Rotation +22° (c=2 in Ethanol)
Boiling Point 174°C to 176°C
UN Number UN2735
Beilstein 1718865
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Amino-1-propanol

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.