Alfa Aesar™ (S)-(+)-2-Amino-1-propanol, 98%

Manufacturer: Alfa Aesar™ B2491603

Quantity 1g

 View more versions of this product

Catalog No. AAB2491603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2749-11-3
    Molecular Formula C3H9NO
    Formula Weight 75.11
    MDL Number MFCD00064412
    UN Number UN2735
    Beilstein 1718865
    Synonym H-Ala-ol; L-Alaninol
    Chemical Name or Material (S)-(+)-2-Amino-1-propanol

    Specifications

    Formula Weight 75.11
    UN Number UN2735
    Beilstein 1718865
    Health Hazard 1 Danger
    Quantity 1g
    Flash Point 83°C (181°F)
    Optical Rotation +22° (c=2 in Ethanol)
    Molecular Formula C3H9NO
    MDL Number MFCD00064412
    CAS 2749-11-3
    Synonym H-Ala-ol; L-Alaninol
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 174° to 176°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (S)-(+)-2-Amino-1-propanol
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit