Alfa Aesar™ (S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+%

Manufacturer: Alfa Aesar™ H52304MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52304MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58577-88-1
    Molecular Formula C10H15NO2
    Formula Weight 181.23
    MDL Number MFCD04112477
    UN Number UN3267
    Chemical Name or Material (S)-(-)-2-Amino-3-benzyloxy-1-propanol

    Specifications

    Formula Weight 181.23
    Molecular Formula C10H15NO2
    MDL Number MFCD04112477
    CAS 58577-88-1
    UN Number UN3267
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Quantity 250mg
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Chemical Name or Material (S)-(-)-2-Amino-3-benzyloxy-1-propanol
    Assay ≥98%

    Description

    Applications
    (S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

    Notes
    Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit