Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Amino-3-methyl-1-butanol, 96%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
2026-48-4
C5H13NO
103.16
MFCD00064296
NWYYWIJOWOLJNR-RXMQYKEDSA-N
l-valinol,s-+-2-amino-3-methyl-1-butanol,2s-2-amino-3-methylbutan-1-ol,l-+-valinol,h-valinol,s-2-amino-3-methylbutan-1-ol,s-2-amino-3-methyl-1-butanol,1-butanol, 2-amino-3-methyl-, 2s,s-2-amino-3-methyl-butan-1-ol
640993
(2S)-2-amino-3-methylbutan-1-ol
CC(C)C(CO)N
2026-48-4
C5H13NO
103.16
MFCD00064296
NWYYWIJOWOLJNR-RXMQYKEDSA-N
l-valinol,s-+-2-amino-3-methyl-1-butanol,2s-2-amino-3-methylbutan-1-ol,l-+-valinol,h-valinol,s-2-amino-3-methylbutan-1-ol,s-2-amino-3-methyl-1-butanol,1-butanol, 2-amino-3-methyl-, 2s,s-2-amino-3-methyl-butan-1-ol
640993
(2S)-2-amino-3-methylbutan-1-ol
CC(C)C(CO)N
Specific Gravity 0.92
Specific Rotation + 16.00
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25mL
Melting Point 30.0°C to 32.0°C
Flash Point 91°C
Color Yellow
Boiling Point 81.0°C (8.0 mmHg)
Refractive Index 1.4538 to 1.4558
Density 0.9200g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula (CH3)2CHCH(NH2)CH2OH
Fieser 16,38
Formula Weight 103.16
Physical Form Liquid After Melting
Percent Purity 96%
Chemical Name or Material (S)-(+)-2-Amino-3-methyl-1-butanol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 217-975-5