Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Amino-4-phenylbutyric acid ethyl ester hydrochloride, 97%, Thermo Scientific™

Catalog Number AC365670050
Click to view available options
5g; Glass bottle
CAS 90891-21-7
CAS Min % 96.0
CAS Max % 100.0
Elemental Analysis 56.76 to 61.48% C, 5.52 to 5.98% N
Loss on Drying 1% max. ()
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C12H17NO2·HCl
Quantity 5g
MDL Number MFCD00190691
Specific Rotation 39.7
Synonym ethyl s-2-amino-4-phenylbutanoate hydrochloride, l-homophenylalanine ethyl ester hydrochloride, h-hophe-oet.hcl, ethyl l-homoalaninate hcl, s-ethyl 2-amino-4-phenylbutanoate hydrochloride, ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, pubchem6054, h-hph-oet.hcl, h-hph-oet. hcl, homophenylalanine ethyl ester hydrochloride
InChI Key PTFKZMFFSIYCOV-MERQFXBCSA-N
SMILES CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
IUPAC Name ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
Molecular Weight (g/mol) 243.73
PubChem CID 2734606
Formula Weight 243.73
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Amino-4-phenylbutyric acid ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit