Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
261360-75-2
C11H15NO3
209.245
HFAZTHPBSQNAFO-VWMHFEHESA-N
2s-2-aminobut-3-en-1-ol; benzoic acid,s-2-aminobut-3-en-1-ol, benzoate salt,s-2-aminobut-3-en-1-ol benzoate,s-2-aminobut-3-en-1-olbenzoate salt,benzoic acid, 2s-2-aminobut-3-en-1-ol
2734532
(2S)-2-aminobut-3-en-1-ol;benzoic acid
C=CC(CO)N.C1=CC=C(C=C1)C(=O)O
261360-75-2
C11H15NO3
209.245
HFAZTHPBSQNAFO-VWMHFEHESA-N
2s-2-aminobut-3-en-1-ol; benzoic acid,s-2-aminobut-3-en-1-ol, benzoate salt,s-2-aminobut-3-en-1-ol benzoate,s-2-aminobut-3-en-1-olbenzoate salt,benzoic acid, 2s-2-aminobut-3-en-1-ol
2734532
(2S)-2-aminobut-3-en-1-ol;benzoic acid
C=CC(CO)N.C1=CC=C(C=C1)C(=O)O
Color White
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 209.24
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (S)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, (98% e.e.)
Show More Show Less