Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
22795-99-9
C7H16N2
128.22
MFCD00191371
UNRBEYYLYRXYCG-ZETCQYMHSA-N
s-1-ethylpyrrolidin-2-yl methanamine,s---2-aminomethyl-1-ethylpyrrolidine,s-2-aminomethyl-1-ethylpyrrolidine,2s-1-ethylpyrrolidin-2-yl methanamine,s-1-ethyl-2-aminomethylpyrrolidine,2s-2-aminomethyl-1-ethylpyrrolidine,2-pyrrolidinemethanamine, 1-ethyl-, 2s,pubchem18590
643457
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
CCN1CCCC1CN
22795-99-9
C7H16N2
128.22
MFCD00191371
UNRBEYYLYRXYCG-ZETCQYMHSA-N
s-1-ethylpyrrolidin-2-yl methanamine,s---2-aminomethyl-1-ethylpyrrolidine,s-2-aminomethyl-1-ethylpyrrolidine,2s-1-ethylpyrrolidin-2-yl methanamine,s-1-ethyl-2-aminomethylpyrrolidine,2s-2-aminomethyl-1-ethylpyrrolidine,2-pyrrolidinemethanamine, 1-ethyl-, 2s,pubchem18590
643457
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
CCN1CCCC1CN
Packaging Glass bottle
Specific Rotation Condition − 107.00 (20.00°C c=1,methanol)
Specific Gravity 0.919
Quantity 1g
Infrared Spectrum Authentic
Specific Rotation − 107.00
Color Colorless to Yellow
Refractive Index 1.4660 to 1.4680
Flash Point 57°C
Density 0.9190g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 128.22
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger