Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
22795-99-9
C7H16N2
128.22
MFCD00191371
UNRBEYYLYRXYCG-ZETCQYMHSA-N
s-1-ethylpyrrolidin-2-yl methanamine,s---2-aminomethyl-1-ethylpyrrolidine,s-2-aminomethyl-1-ethylpyrrolidine,2s-1-ethylpyrrolidin-2-yl methanamine,s-1-ethyl-2-aminomethylpyrrolidine,2s-2-aminomethyl-1-ethylpyrrolidine,2-pyrrolidinemethanamine, 1-ethyl-, 2s,pubchem18590
643457
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
CCN1CCCC1CN
22795-99-9
C7H16N2
128.22
MFCD00191371
UNRBEYYLYRXYCG-ZETCQYMHSA-N
s-1-ethylpyrrolidin-2-yl methanamine,s---2-aminomethyl-1-ethylpyrrolidine,s-2-aminomethyl-1-ethylpyrrolidine,2s-1-ethylpyrrolidin-2-yl methanamine,s-1-ethyl-2-aminomethylpyrrolidine,2s-2-aminomethyl-1-ethylpyrrolidine,2-pyrrolidinemethanamine, 1-ethyl-, 2s,pubchem18590
643457
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
CCN1CCCC1CN
Specific Gravity 0.919
Packaging Glass bottle
Quantity 25g
Refractive Index 1.4660 to 1.4680
Infrared Spectrum Authentic
Density 0.9190g/mL
Specific Rotation − 107.00
Specific Rotation Condition − 107.00 (20.00°C c=1,methanol)
Color Colorless to Yellow
Flash Point 57°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 128.22
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger