Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-(Aminomethyl)pyrrolidine, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
69500-64-7
C5H12N2
100.165
MFCD00191745
AUKXFNABVHIUAC-YFKPBYRVSA-N
s-pyrrolidin-2-ylmethanamine,s-+-2-aminomethyl pyrrolidine,s-2-aminomethyl pyrrolidine,2s-pyrrolidin-2-ylmethylamine,s-pyrrolidin-2-ylmethylamine,s-2-aminomethylpyrrolidine,2s-pyrrolidin-2-yl methanamine,2-pyrrolidinemethanamine, 2s,1-2s-pyrrolidin-2-yl methanamine,d07vdj
2734054
CHEBI:44632
[(2S)-pyrrolidin-2-yl]methanamine
C1CC(NC1)CN
69500-64-7
C5H12N2
100.165
MFCD00191745
AUKXFNABVHIUAC-YFKPBYRVSA-N
s-pyrrolidin-2-ylmethanamine,s-+-2-aminomethyl pyrrolidine,s-2-aminomethyl pyrrolidine,2s-pyrrolidin-2-ylmethylamine,s-pyrrolidin-2-ylmethylamine,s-2-aminomethylpyrrolidine,2s-pyrrolidin-2-yl methanamine,2-pyrrolidinemethanamine, 2s,1-2s-pyrrolidin-2-yl methanamine,d07vdj
2734054
CHEBI:44632
[(2S)-pyrrolidin-2-yl]methanamine
C1CC(NC1)CN
Quantity 250mg
Density 0.933g/mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 100.16
Chemical Name or Material (S)-(+)-2-(Aminomethyl)pyrrolidine

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger