Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Benzylsuccinic acid-1-methyl ester, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
182247-45-6
C12H14O4
222.24
MFCD01311761
BUNMUVFKMIOEQU-JTQLQIEISA-N
s-3-benzyl-4-methoxy-4-oxobutanoic acid,s---2-benzylsuccinic acid 1-methyl ester,3s-3-benzyl-4-methoxy-4-oxobutanoic acid,s-2-benzylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-4-phenylbutanoic acid,butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
7006556
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
182247-45-6
C12H14O4
222.24
MFCD01311761
BUNMUVFKMIOEQU-JTQLQIEISA-N
s-3-benzyl-4-methoxy-4-oxobutanoic acid,s---2-benzylsuccinic acid 1-methyl ester,3s-3-benzyl-4-methoxy-4-oxobutanoic acid,s-2-benzylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-4-phenylbutanoic acid,butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
7006556
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
Color Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 222.24
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material (S)-2-Benzylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less