Alfa Aesar™ (S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%

Manufacturer: Alfa Aesar™ H5196406

Quantity 5g

 View more versions of this product

Catalog No. AAH5196406

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 122235-70-5
    Molecular Formula C23H26N2O6
    Formula Weight 426.47
    MDL Number MFCD00235897
    Beilstein 4771385
    Synonym Boc-Dap(Fmoc)-OH
    Chemical Name or Material (S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid

    Specifications

    Formula Weight 426.47
    Molecular Formula C23H26N2O6
    MDL Number MFCD00235897
    CAS 122235-70-5
    Synonym Boc-Dap(Fmoc)-OH
    Quantity 5g
    Beilstein 4771385
    Chemical Name or Material (S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid
    Assay 98%

    Description

    Applications
    It has been used in the synthesis of target chelator, the tris-catechol compound 2.

    Solubility
    Soluble in 2ml dimethyl formamide 0.3gram.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit