Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%

Manufacturer:  Alfa Aesar™ H5196403

Quantity 1g

 View more versions of this product

Catalog No. AAH5196403


  Check Availability
Add to cart

Specifications

Description

Applications
It has been used in the synthesis of target chelator, the tris-catechol compound 2.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

122235-70-5
426.47
4771385
(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid
C23H26N2O6
MFCD00235897
Boc-Dap(Fmoc)-OH

Specifications

122235-70-5
426.47
4771385
1g
98%
C23H26N2O6
MFCD00235897
Boc-Dap(Fmoc)-OH
(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit