Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-chloromandelic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
52950-19-3
C8H7ClO3
186.591
MFCD00798437
RWOLDZZTBNYTMS-ZETCQYMHSA-N
s-+-2-chloromandelic acid,s-2-2-chlorophenyl-2-hydroxyacetic acid,s-2-chloromandelic acid,s-o-chloromandelicacid,2-chloro-l-mandelic acid,2s-2-2-chlorophenyl-2-hydroxy-acetic acid,s-2-chlorophenyl hydroxy acetic acid,pubchem5743,s-o-chloromandelic acid,ksc914s2j
6922874
(2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
C1=CC=C(C(=C1)C(C(=O)O)O)Cl
52950-19-3
C8H7ClO3
186.591
MFCD00798437
RWOLDZZTBNYTMS-ZETCQYMHSA-N
s-+-2-chloromandelic acid,s-2-2-chlorophenyl-2-hydroxyacetic acid,s-2-chloromandelic acid,s-o-chloromandelicacid,2-chloro-l-mandelic acid,2s-2-2-chlorophenyl-2-hydroxy-acetic acid,s-2-chlorophenyl hydroxy acetic acid,pubchem5743,s-o-chloromandelic acid,ksc914s2j
6922874
(2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
C1=CC=C(C(=C1)C(C(=O)O)O)Cl
Melting Point 118°C to 122°C
Infrared Spectrum Authentic
Quantity 1g
Color White to Brown
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H4CH2(OH)COOH
Formula Weight 186.59
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material (S)-(+)-2-Chloromandelic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.