Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
220497-69-8
C12H20O4
228.288
MFCD01311762
TYYNOKUMAOAVBK-JTQLQIEISA-N
s-2-cyclohexylmethyl succinic acid-1-methyl ester,s-4-methoxy-3-cyclohexylmethyl-4-oxobutanoic acid,s-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid,s-2-cyclohexylmethyl-succinic acid 1-methyl ester,s-2-cyclohexylmethyl succinic acid 1-methyl ester,3s-4-cyclohexyl-3-methoxycarbonyl butanoic acid,butanedioic acid,2-cyclohexylmethyl-, 1-methyl ester, 2s,pubchem23240,s-3-methoxycarbonyl-4-cyclohexylbutyric acid,s-cyclohexylmethylsuccinic acid 1-methyl ester
22831490
(3S)-3-(cyclohexylmethyl)-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1CCCCC1)CC(=O)O
220497-69-8
C12H20O4
228.288
MFCD01311762
TYYNOKUMAOAVBK-JTQLQIEISA-N
s-2-cyclohexylmethyl succinic acid-1-methyl ester,s-4-methoxy-3-cyclohexylmethyl-4-oxobutanoic acid,s-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid,s-2-cyclohexylmethyl-succinic acid 1-methyl ester,s-2-cyclohexylmethyl succinic acid 1-methyl ester,3s-4-cyclohexyl-3-methoxycarbonyl butanoic acid,butanedioic acid,2-cyclohexylmethyl-, 1-methyl ester, 2s,pubchem23240,s-3-methoxycarbonyl-4-cyclohexylbutyric acid,s-cyclohexylmethylsuccinic acid 1-methyl ester
22831490
(3S)-3-(cyclohexylmethyl)-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1CCCCC1)CC(=O)O
Color Colorless to Yellow
Quantity 1g
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 228.28
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material (S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less