Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Diphenylmethylpyrrolidine, 97+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
119237-64-8
C17H19N
237.34
MFCD00799525
OXOBKZZXZVFOBB-INIZCTEOSA-N
s-2-benzhydrylpyrrolidine,s---2-diphenylmethyl pyrrolidine,s-2-diphenylmethylpyrrolidine,2s-2-diphenylmethyl pyrrolidine,2s-2-benzhydrylpyrrolidine,2s-2-benzylhydrylpyrrolidine,2r-2-benzhydrylpyrrolidine,pubchem18168,2alpha-benzhydrylpyrrolidine,s-2-benzhydryl-pyrrolidine
2734053
(2S)-2-benzhydrylpyrrolidine
C1CC(NC1)C(C2=CC=CC=C2)C3=CC=CC=C3
119237-64-8
C17H19N
237.34
MFCD00799525
OXOBKZZXZVFOBB-INIZCTEOSA-N
s-2-benzhydrylpyrrolidine,s---2-diphenylmethyl pyrrolidine,s-2-diphenylmethylpyrrolidine,2s-2-diphenylmethyl pyrrolidine,2s-2-benzhydrylpyrrolidine,2s-2-benzylhydrylpyrrolidine,2r-2-benzhydrylpyrrolidine,pubchem18168,2alpha-benzhydrylpyrrolidine,s-2-benzhydryl-pyrrolidine
2734053
(2S)-2-benzhydrylpyrrolidine
C1CC(NC1)C(C2=CC=CC=C2)C3=CC=CC=C3
Specific Rotation Condition - 3.00 (25.00°C C=1,Chloroform)
Color Colorless to Yellow
Flash Point 109°C
Infrared Spectrum Authentic
Refractive Index 1.5860 to 1.5880
Packaging Glass bottle
Density 1.0620g/mL
Quantity 1g
Specific Rotation − 3.00
Specific Gravity 1.062
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 237.34
Physical Form Oil
Percent Purity ≥97%
Chemical Name or Material (S)-2-Diphenylmethylpyrrolidine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.