Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-2-(Fmoc-amino)-4-aminobutyric acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
161420-87-7
C19H20N2O4
340.379
MFCD00237017
ZZDRDGKSMGGBDI-KRWDZBQOSA-N
fmoc-dab-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid,fmoc-l-2,4-diaminobutyric acid,2s-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,s-n2-fmoc-2,4-diaminobutyric acid,ambotzfaa1456,na-fmoc-l-2,4-diaminobutyric acid,n-a-fmoc-l-2,4-diaminobutyric acid,2s-2-9h-fluorene-9-ylmethoxycarbonylamino-4-aminobutanoic acid,2s-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
7019708
(2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
161420-87-7
C19H20N2O4
340.379
MFCD00237017
ZZDRDGKSMGGBDI-KRWDZBQOSA-N
fmoc-dab-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid,fmoc-l-2,4-diaminobutyric acid,2s-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,s-n2-fmoc-2,4-diaminobutyric acid,ambotzfaa1456,na-fmoc-l-2,4-diaminobutyric acid,n-a-fmoc-l-2,4-diaminobutyric acid,2s-2-9h-fluorene-9-ylmethoxycarbonylamino-4-aminobutanoic acid,2s-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
7019708
(2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
Color White
Quantity 250mg
Formula Weight 340.38
Physical Form Powder
Percent Purity 98%
Chemical Name or Material (S)-2-(Fmoc-amino)-4-aminobutyric acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C