Alfa Aesar™ (S)-(+)-2-Heptanol, 98%

Manufacturer: Alfa Aesar™ 3379503

Quantity 1g

 View more versions of this product

Catalog No. AA3379503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6033-23-4
    Formula Weight 116.2
    MDL Number MFCD00065003
    UN Number UN1987
    Merck Index 14,4662
    Chemical Name or Material (S)-(+)-2-Heptanol

    Specifications

    Formula Weight 116.2
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1987
    Merck Index 14,4662
    Density 0.815g/mL
    MDL Number MFCD00065003
    Refractive Index 1.4210
    Quantity 1g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Physical Form Liquid
    Health Hazard 1 Danger
    Boiling Point 160° to 162°C
    Flash Point 59° (138°F)
    CAS 6033-23-4
    Health Hazard 2 GHS H Statement
    H311-H226-H319
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Causes serious eye irritation.
    Chemical Name or Material (S)-(+)-2-Heptanol
    Assay 98%

    GHS H Statement
    H311-H226-H319
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Causes serious eye irritation.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit