Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass Bottle
213270-36-1
C9H16O4
188.22
MFCD01091033
AJNNFMRCMKGYHA-ZETCQYMHSA-N
s-3-methoxycarbonyl-5-methylhexanoic acid,s---2-isobutylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-5-methylhexanoic acid,s-2-isobutylsuccinic acid 1-methyl ester,s-2-2-methylpropyl butanedioic acid 1-methyl ester,butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
2734513
(3S)-3-methoxycarbonyl-5-methylhexanoic acid
CC(C)CC(CC(=O)O)C(=O)OC
213270-36-1
C9H16O4
188.22
MFCD01091033
AJNNFMRCMKGYHA-ZETCQYMHSA-N
s-3-methoxycarbonyl-5-methylhexanoic acid,s---2-isobutylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-5-methylhexanoic acid,s-2-isobutylsuccinic acid 1-methyl ester,s-2-2-methylpropyl butanedioic acid 1-methyl ester,butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
2734513
(3S)-3-methoxycarbonyl-5-methylhexanoic acid
CC(C)CC(CC(=O)O)C(=O)OC
Packaging Glass Bottle
Quantity 1g
Formula Weight 188.22
Percent Purity 95%,(98% ee)
Chemical Name or Material (S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove

GHS Signal Word: Warning

TSCA : TSCA