Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
208113-95-5
C8H14O4
174.19
MFCD01091031
VHOGSWBAPDPWTG-LURJTMIESA-N
s-2-isopropylsuccinic acid-1-methyl ester,s-3-methoxycarbonyl-4-methylpentanoic acid,s-2-isopropylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-4-methylpentanoic acid,butanedioic acid, 2-1-methylethyl-, 1-methyl ester, 2s,s-2-isopropylsuccinic acid-1-methylester,2-s-isopropylsuccinic acid 1-methyl ester,s-+-2-isopropylsuccinic acid 1-methyl ester
2734514
(3S)-3-methoxycarbonyl-4-methylpentanoic acid
CC(C)C(CC(=O)O)C(=O)OC
208113-95-5
C8H14O4
174.19
MFCD01091031
VHOGSWBAPDPWTG-LURJTMIESA-N
s-2-isopropylsuccinic acid-1-methyl ester,s-3-methoxycarbonyl-4-methylpentanoic acid,s-2-isopropylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-4-methylpentanoic acid,butanedioic acid, 2-1-methylethyl-, 1-methyl ester, 2s,s-2-isopropylsuccinic acid-1-methylester,2-s-isopropylsuccinic acid 1-methyl ester,s-+-2-isopropylsuccinic acid 1-methyl ester
2734514
(3S)-3-methoxycarbonyl-4-methylpentanoic acid
CC(C)C(CC(=O)O)C(=O)OC
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 174.19
Physical Form Viscous Liquid
Percent Purity 95%,98% ee
Chemical Name or Material (S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less