Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Methoxypropionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
2.5g
Packaging:
Glass bottle
23953-00-6
C4H8O3
104.11
MFCD01632589
ICPWFHKNYYRBSZ-VKHMYHEASA-N
s-2-methoxypropanoic acid,s---2-methoxypropionic acid,2s-2-methoxypropanoic acid,propanoic acid, 2-methoxy-, 2s,unii-74k293ocx6,2-methoxypropanoic acid, s,propanoic acid,2-methoxy-, 2s,propanoic acid, 2-methoxy-, s,propionic acid, 2-methoxy-, s--,s-2-methoxypropionic acid
5702566
(2S)-2-methoxypropanoic acid
CC(C(=O)O)OC
23953-00-6
C4H8O3
104.11
MFCD01632589
ICPWFHKNYYRBSZ-VKHMYHEASA-N
s-2-methoxypropanoic acid,s---2-methoxypropionic acid,2s-2-methoxypropanoic acid,propanoic acid, 2-methoxy-, 2s,unii-74k293ocx6,2-methoxypropanoic acid, s,propanoic acid,2-methoxy-, 2s,propanoic acid, 2-methoxy-, s,propionic acid, 2-methoxy-, s--,s-2-methoxypropionic acid
5702566
(2S)-2-methoxypropanoic acid
CC(C(=O)O)OC
Specific Rotation Condition −75.00 (20.00°C c=neat)
Infrared Spectrum Authentic
Refractive Index 1.4132 to 1.4152
Specific Rotation − 75.00
Packaging Glass bottle
Quantity 2.5g
Color Yellow
Flash Point 87°C
Boiling Point 99.0°C (20.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH(OCH3)COOH
Beilstein 03, III, 444
Formula Weight 104.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (S)-(-)-2-Methoxypropionic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA