Alfa Aesar™ (S)-(-)-2-Methyl-2-propanesulfinamide, 97%

Manufacturer: Alfa Aesar™ H2711506

Quantity 5g

 View more versions of this product

Catalog No. AAH2711506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 343338-28-3
    Molecular Formula C4H11NOS
    Formula Weight 121.2
    MDL Number MFCD05861480
    Synonym (S)-(-)-tert-Butyl sulfinamide
    Chemical Name or Material (S)-(-)-2-Methyl-2-propanesulfinamide

    Specifications

    Formula Weight 121.2
    Molecular Formula C4H11NOS
    MDL Number MFCD05861480
    CAS 343338-28-3
    Synonym (S)-(-)-tert-Butyl sulfinamide
    Melting Point 97° to 101°C
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material (S)-(-)-2-Methyl-2-propanesulfinamide
    Assay 97%

    Description

    Applications
    (S)-(-)-2-Methyl-2-propanesulfinamide is used in Suzuki reaction. It is also employed as a reagent for synthesizing chiral amines. It acts as a chiral auxiliary used in an asymmetric synthesis of trifluoroethylamines by conversion of trifluoroacetaldehyde to a chiral imine. It is also involved in the transformation of P,N-sulfinyl imine ligands through condensation with aldehydes and ketones, which can undergo iridium-catalyzed asymmetric hydrogenation of olefins. Further, it serves as a reagent for the preparation of chemicals and pharmaceutical intermediates.

    Notes
    Incompatible with strong oxidizing agents. Store in a cool place.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit