Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
112022-81-8,108-88-3
C18H20BNO
277.17
MFCD00078439
VMKAFJQFKBASMU-KRWDZBQOSA-N
s-2-methyl-cbs-oxazaborolidine,s-methyl oxazaborolidine,s---2-methyl-cbs-oxazaborolidine,s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine,corey catalyst,s-methyl-cbs-oxazaborolidine,s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole,s-me-cbs catalyst,s-methyl-cbs,s-me-cbs
2734713
(3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C
112022-81-8,108-88-3
C18H20BNO
277.17
MFCD00078439
VMKAFJQFKBASMU-KRWDZBQOSA-N
s-2-methyl-cbs-oxazaborolidine,s-methyl oxazaborolidine,s---2-methyl-cbs-oxazaborolidine,s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine,corey catalyst,s-methyl-cbs-oxazaborolidine,s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole,s-me-cbs catalyst,s-methyl-cbs,s-me-cbs
2734713
(3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C
Flash Point 4°C
Appearance Colorless to amber solution
Packaging Glass bottle
Boiling Point 111.0°C
Specific Gravity 0.929
Density 0.9290g/mL
Quantity 25mL
CAS Min % 28.34,67.18
CAS Max % 32.82,71.66
Solubility Information Solubility in water: reacts
Formula Weight 277.17
Chemical Name or Material (S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Show More Show Less

GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging the unborn child.
May cause damage to organs through prolong

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov

GHS Signal Word: Danger