Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methylpiperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
74879-18-8
C5H12N2
100.16
MFCD00171452
JOMNTHCQHJPVAZ-YFKPBYRVSA-N
s-+-2-methylpiperazine,s-2-methylpiperazine,2s-2-methylpiperazine,piperazine, 2-methyl-, 2s,s-2-methyl-piperazine,unii-c5vi3s1yc0,2-methylpiperazine, s,2-s-methylpiperazine,s-mp,c5vi3s1yc0
2734219
(2S)-2-methylpiperazine
CC1CNCCN1
74879-18-8
C5H12N2
100.16
MFCD00171452
JOMNTHCQHJPVAZ-YFKPBYRVSA-N
s-+-2-methylpiperazine,s-2-methylpiperazine,2s-2-methylpiperazine,piperazine, 2-methyl-, 2s,s-2-methyl-piperazine,unii-c5vi3s1yc0,2-methylpiperazine, s,2-s-methylpiperazine,s-mp,c5vi3s1yc0
2734219
(2S)-2-methylpiperazine
CC1CNCCN1
Quantity 5g
Color White to Yellow
Melting Point 86°C to 91°C
Packaging Glass bottle
Specific Rotation + 13.5
Boiling Point 155°C to 156°C
Flash Point 65°C
Specific Rotation Condition + 13.50 (20.00°C c=1,toluene)
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: soluble
Formula Weight 100.16
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Methylpiperazine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA