Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
98-79-3
C5H7NO3
129.12
ODHCTXKNWHHXJC-VKHMYHEASA-N
l-pyroglutamic acid,pyroglutamic acid,pidolic acid,5-oxo-l-proline,h-pyr-oh,5-oxoproline,s-5-oxopyrrolidine-2-carboxylic acid,2s-5-oxopyrrolidine-2-carboxylic acid,l-5-oxoproline,l-proline, 5-oxo
7405
CHEBI:18183
(2S)-5-oxopyrrolidine-2-carboxylic acid
C1CC(=O)NC1C(=O)O
98-79-3
C5H7NO3
129.12
ODHCTXKNWHHXJC-VKHMYHEASA-N
l-pyroglutamic acid,pyroglutamic acid,pidolic acid,5-oxo-l-proline,h-pyr-oh,5-oxoproline,s-5-oxopyrrolidine-2-carboxylic acid,2s-5-oxopyrrolidine-2-carboxylic acid,l-5-oxoproline,l-proline, 5-oxo
7405
CHEBI:18183
(2S)-5-oxopyrrolidine-2-carboxylic acid
C1CC(=O)NC1C(=O)O
Specific Rotation Condition − 27.50 (20.00°C c=10,1 N NaOH)
Specific Rotation − 27.50
Water 0.5% Max. (K.F.)
Quantity 25g
Packaging Plastic bottle
Infrared Spectrum Authentic
Melting Point 152.0°C to 162.0°C
Color White
Assay Percent Range 98%
Beilstein 22,284
Merck Index 15,8113
Solubility Information Solubility in water: 100-150g/l (20°). Other solubilities: soluble in alcohol and acetone
Formula Weight 129.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (S)-(-)-2-Pyrrolidone-5-carboxylic acid
Show More Show Less

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 202-700-3